Learn More
ACE-2 Goat anti-Human, Alexa Fluor 700, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems FAB933N100UG
Catalog No. FAB933N100U
Add to cart
This item is not returnable. View return policy
Description
Description
ACE-2 Polyclonal antibody specifically detects ACE-2 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
ACE-2
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Goat
IgG
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Polyclonal
Human
Flow Cytometry
Flow Cytometry 0.25-1 ug/10^6 cells
Q9BYF1
ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Mouse myeloma cell line NS0-derived recombinant human ACE-2, Gln18-Ser740, Accession # Q9BYF1
Antigen Affinity-purified
RUO
Primary
59272