Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACE-2, Mouse anti-Human, Clone: AC18F, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
$350.00 - $482.50
Specifications
Antigen | ACE-2 |
---|---|
Clone | AC18F |
Concentration | 1 mg/mL |
Dilution | Western Blot, Flow Cytometry, ELISA |
Applications | Western Blot, Flow Cytometry, ELISA |
Description
ACE-2 Monoclonal specifically detects ACE-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
ACE-2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Immunology, Virology Bacteria and Parasites | |
0.2 um-filtered solution in PBS, pH 7.4 | |
59272 | |
Recombinant human ACE-2. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
AC18F | |
Western Blot, Flow Cytometry, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
ACE2 | |
IgG1 κ | |
Protein G purified | |
Recognizes human ACE-2. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title