Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACE-2 Mouse anti-Human, Hamster, PerCP, Clone: AC384, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP280038PCP

Catalog No. NB065787


Add to cart

Description

Description

ACE-2 Monoclonal antibody specifically detects ACE-2 in Human, Hamster samples. It is validated for Flow Cytometry
Specifications

Specifications

ACE-2
AC384
PerCP
PBS
ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Recombinant human ACE-2.
Protein G purified
RUO
Store at 4°C in the dark.
Monoclonal
Hamster, Human
Flow Cytometry
1 mg/ml
Flow Cytometry
Purified
Mouse
IgG1 Kappa
0.1 mL
Immunology, Virology Bacteria and Parasites
Primary
59272
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit