Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACE-2 Rat anti-Human, Alexa Fluor 350, Clone: 379131, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB9334U100UG
This item is not returnable.
View return policy
Description
ACE-2 Monoclonal specifically detects ACE-2 in Human samples. It is validated for Flow Cytometry.Specifications
ACE-2 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
Mouse myeloma cell line, NS0-derived human ACE-2, Gln18-Ser740, Accession # Q9BYF1 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
379131 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9BYF1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
59272 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title