Learn More
ACE/CD143 Mouse anti-Human, Alexa Fluor 594, Clone: 171417, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB929T100UG
Catalog No. FAB929T100
Add to cart
This item is not returnable. View return policy
Description
Description
ACE/CD143 Monoclonal antibody specifically detects ACE/CD143 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
ACE/CD143
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA
Mouse
IgG1
RUO
Primary
1636
Human
Flow Cytometry
171417
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
ACE
Mouse myeloma cell line NS0-derived recombinant human ACE/CD143 aa 30-1261
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human ACE/CD143 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ACE-2 is observed. Detects the surface expression of human ACE on full length ACE transfectants, but not on control transfectants by flow cytometry.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)