Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACE/CD143 Mouse anti-Human, Clone: ACE/3763, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30760820UG
Description
ACE/CD143 Monoclonal specifically detects ACE/CD143 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.Specifications
ACE/CD143 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml | |
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
Recombinant fragment (around aa 652-788) of human ACE/CD143 protein | |
20 μg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
ACE/3763 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
1636 | |
Store at 4°C. | |
IgG2c κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title