Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACE/CD143 Rabbit anti-Human, Alexa Fluor 350, Clone: ACE/7004R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314036AF350
This item is not returnable.
View return policy
Description
ACE/CD143 Monoclonal antibody specifically detects ACE/CD143 in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
ACE/CD143 | |
Monoclonal | |
Alexa Fluor 350 | |
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
Recombinant fragment (around aa 652-788) of human ACE/CD143 protein (Uniprot: P12821) | |
0.1 mL | |
Cancer, Cardiovascular Biology, CD Markers, Cell Biology, Endocrinology, Immunology, Signal Transduction, Stem Cells | |
1636 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
ACE/7004R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title