Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC AQ 2UM 100X2.1MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1061002U
ACE Excel AQ, 2um, Ultra-Inert UHPLC Column, 100x2.1mm
Catalog No. 50-147-654