Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C18 2UM 20X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1010203U
ACE Excel C18, 2um, Ultra-Inert UHPLC Column, 20x3.0mm
Catalog No. 50-147-520