Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C18 5UM 30X4.6MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1210346U
ACE Excel C18, 5um, Ultra-Inert UHPLC Column, 30x4.6mm
Catalog No. 50-147-953