Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C18-AR 2UM 50X4.6MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1090546U
ACE Excel C18-AR, 2um, Ultra-Inert UHPLC Column, 50x4.6mm
Catalog No. 50-147-701