Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C18-AR 3UM 125X4.6MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1191246U
ACE Excel C18-AR, 3um, Ultra-Inert UHPLC Column, 125x4.6mm
Catalog No. 50-147-914