Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C18-AR 5UM 100X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1291003U
ACE Excel C18-AR, 5um, Ultra-Inert UHPLC Column, 100x3.0mm
Catalog No. 50-148-144