Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C18-PFP 3UM 30X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL11100303U
ACE Excel C18-PFP, 3um, Ultra-Inert UHPLC Column, 30x3.0mm
Catalog No. 50-147-721