Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C18PFP 5UM 300X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL12103003U
ACE Excel C18-PFP, 5um, Ultra-Inert UHPLC Column, 300x3.0mm
Catalog No. 50-147-949