Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C4 2UM 100X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1031003U
ACE Excel C4, 2um, Ultra-Inert UHPLC Column, 100x3.0mm
Catalog No. 50-147-583