Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C4 2UM 150X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1031503U
ACE Excel C4, 2um, Ultra-Inert UHPLC Column, 150x3.0mm
Catalog No. 50-147-589