Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C4 3UM 125X4.6MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1131246U
ACE Excel C4, 3um, Ultra-Inert UHPLC Column, 125x4.6mm
Catalog No. 50-147-806