Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C4 3UM 50X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1130503U
ACE Excel C4, 3um, Ultra-Inert UHPLC Column, 50x3.0mm
Catalog No. 50-147-799