Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C4 5UM 20X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1230203U
ACE Excel C4, 5um, Ultra-Inert UHPLC Column, 20x3.0mm
Catalog No. 50-148-015