Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C4 5UM 300X2.1MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1233002U
ACE Excel C4, 5um, Ultra-Inert UHPLC Column, 300x2.1mm
Catalog No. 50-148-035