Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC C8 5UM 250X2.1MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1222502U
ACE Excel C8, 5um, Ultra-Inert UHPLC Column, 250x2.1mm
Catalog No. 50-148-002