Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC CN 2UM 150X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1041503U
ACE Excel CN, 2um, Ultra-Inert UHPLC Column, 150x3.0mm
Catalog No. 50-147-613