Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC CN 3UM 100X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1141003U
ACE Excel CN, 3um, Ultra-Inert UHPLC Column, 100x3.0mm
Catalog No. 50-147-829