Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC CN 3UM 35X2.1MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1143502U
ACE Excel CN, 3um, Ultra-Inert UHPLC Column, 35x2.1mm
Catalog No. 50-147-840