Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC CN 5UM 125X2.1MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1241202U
ACE Excel CN, 5um, Ultra-Inert UHPLC Column, 125x2.1mm
Catalog No. 50-148-056