Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC CN 5UM 75X2.1MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1247502U
ACE Excel CN, 5um, Ultra-Inert UHPLC Column, 75x2.1mm
Catalog No. 50-148-071