Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC PHENYL 5UM 150X3.0MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1251503U
ACE Excel Phenyl, 5um, Ultra-Inert UHPLC Column, 150x3.0mm
Catalog No. 50-148-090