Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC PHENYL 5UM 35X4.6MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1253546U
ACE Excel Phenyl, 5um, Ultra-Inert UHPLC Column, 35x4.6mm
Catalog No. 50-148-100