Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC SIL 2UM 30X2.1MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1070302U
ACE Excel SIL, 2um, Ultra-Inert UHPLC Column, 30x2.1mm
Catalog No. 50-147-672