Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC SIL 2UM 75X2.1MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1077502U
ACE Excel SIL, 2um, Ultra-Inert UHPLC Column, 75x2.1mm
Catalog No. 50-147-690