Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC SIL 3UM 50X2.1MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1170502U
ACE Excel SIL, 3um, Ultra-Inert UHPLC Column, 50x2.1mm
Catalog No. 50-147-879