Learn More
MAC-MOD AnalyticalSupplier Diversity Partner ACE UHPLC SIL 5UM 30X4.6MM  DFS Item
Manufacturer:  MAC-MOD AnalyticalSupplier Diversity Partner EXL1270346U
ACE Excel SIL, 5um, Ultra-Inert UHPLC Column, 30x4.6mm
Catalog No. 50-148-109