Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACE2 Monoclonal Antibody (OTI4D2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526628
Description
ACE2 Monoclonal antibody specifically detects ACE2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ACE2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BYF1 | |
ACE2 | |
Human recombinant protein fragment corresponding to amino acids 18-237 of ACE2 produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4D2 | |
Unconjugated | |
ACE2 | |
2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885 | |
Mouse | |
Affinity Chromatography | |
RUO | |
59272 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction