Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACEA, Tocris Bioscience™

Click to view available options
Quantity:
5mg
25mg
This item is not returnable. View return policy
220556-69-4
C22H36ClNO
365.986
SCJNCDSAIRBRIA-CGRWFSSPSA-N
arachidonyl-2 inverted exclamation marka-chloroethylamide hydrate,5e,8e,11e,14e-n-2-chloroethyl icosa-5,8,11,14-tetraenamide
5353377
(5E,8E,11E,14E)-N-(2-chloroethyl)icosa-5,8,11,14-tetraenamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCCl
This item is not returnable. View return policy
220556-69-4
C22H36ClNO
365.986
SCJNCDSAIRBRIA-CGRWFSSPSA-N
arachidonyl-2 inverted exclamation marka-chloroethylamide hydrate,5e,8e,11e,14e-n-2-chloroethyl icosa-5,8,11,14-tetraenamide
5353377
(5E,8E,11E,14E)-N-(2-chloroethyl)icosa-5,8,11,14-tetraenamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCCl

Potent and highly selective CB1 agonist

Potent and highly selective CB1 receptor agonist (Ki = 1.4 nM). Displays >1400-fold selectivity over CB2 receptors. Active in vivo.
Quantity 5mg
Assay Percent Range >98%
Formula Weight 365.99
Chemical Name or Material ACEA

Recommended Storage : Store at -20°C