Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aceclofenac, 99%

Click to view available options
Quantity:
1g
5g
10g
89796-99-6
C16H13Cl2NO4
354.183
MFCD00864296
MNIPYSSQXLZQLJ-UHFFFAOYSA-N
aceclofenac,aceclofenaco,aceclofenacum,preservex,airtal,falcol,gerbin,aceclofar,aceclofenacum latin,aceclofenaco spanish
71771
CHEBI:31159
2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid
C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl
89796-99-6
C16H13Cl2NO4
354.183
MFCD00864296
MNIPYSSQXLZQLJ-UHFFFAOYSA-N
aceclofenac,aceclofenaco,aceclofenacum,preservex,airtal,falcol,gerbin,aceclofar,aceclofenacum latin,aceclofenaco spanish
71771
CHEBI:31159
2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid
C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl
A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation.
Melting Point 149°C to 153°C
Quantity 1g
Color White
UN Number UN2811
Merck Index 14,22
Solubility Information Soluble in DMSO
Formula Weight 354.18
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Aceclofenac

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : CY1577900