Learn More
Aceclofenac, EP, 99-101%, Spectrum™
A3512, 89796-99-6, C16H13Cl2NO4
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner A35125KGBL
 View more versions of this product
Catalog No. 18-600-162
Add to cart
Description
Description
Spectrum™ Aceclofenac, EP is a non-steroidal anti-inflammatory drug with strong analgesic properties. Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
0.005
0.001
89796-99-6
UN2811
Poly Pail
5 kg
1
99 to 101%