Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acenaphthene, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
83-32-9
C12H10
154.21
MFCD00003807
CWRYPZZKDGJXCA-UHFFFAOYSA-N
acenaphthene,1,8-ethylenenaphthalene,naphthyleneethylene,peri-ethylenenaphthalene,acenaphthylene, 1,2-dihydro,ethylenenaphthalene,1,8-dihydroacenaphthalene,unii-v8ut1gac5y,ccris 5951,v8ut1gac5y
6734
CHEBI:22154
1,2-dihydroacenaphthylene
C1CC2=CC=CC3=C2C1=CC=C3
83-32-9
C12H10
154.21
MFCD00003807
CWRYPZZKDGJXCA-UHFFFAOYSA-N
acenaphthene,1,8-ethylenenaphthalene,naphthyleneethylene,peri-ethylenenaphthalene,acenaphthylene, 1,2-dihydro,ethylenenaphthalene,1,8-dihydroacenaphthalene,unii-v8ut1gac5y,ccris 5951,v8ut1gac5y
6734
CHEBI:22154
1,2-dihydroacenaphthylene
C1CC2=CC=CC3=C2C1=CC=C3
Flash Point 135°C
Boiling Point 279°C
Specific Gravity 1.06
Melting Point 90°C to 95°C
Quantity 500g
Density 1.0600g/mL
Color Brown
Packaging Plastic bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Beilstein 05, IV, 1834
Merck Index 15, 29
Solubility Information Solubility in water: insoluble. Other solubilities: 3.2g/100mL glacial acetic acid,1g/31mL alcohol- 1g/56mL methanol,1g/25mL propanol- 1g/2.5mL chloroform,1g/5mL benzene- 1g/5mL toluene
Formula Weight 154.21
Physical Form Crystalline Powder and Chunks
Percent Purity 99%
Chemical Name or Material Acenaphthene
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 201-469-6