Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acenaphthene Zone Refined (number of passes:30) 99.5+%, TCI America™

Click to view available options
Quantity:
ZONE
83-32-9
C12H10
154.212
MFCD00003807
CWRYPZZKDGJXCA-UHFFFAOYSA-N
acenaphthene,1,8-ethylenenaphthalene,naphthyleneethylene,peri-ethylenenaphthalene,acenaphthylene, 1,2-dihydro,ethylenenaphthalene,1,8-dihydroacenaphthalene,unii-v8ut1gac5y,ccris 5951,v8ut1gac5y
6734
CHEBI:22154
1,2-dihydroacenaphthylene
C1CC2=CC=CC3=C2C1=CC=C3
83-32-9
C12H10
154.212
MFCD00003807
CWRYPZZKDGJXCA-UHFFFAOYSA-N
acenaphthene,1,8-ethylenenaphthalene,naphthyleneethylene,peri-ethylenenaphthalene,acenaphthylene, 1,2-dihydro,ethylenenaphthalene,1,8-dihydroacenaphthalene,unii-v8ut1gac5y,ccris 5951,v8ut1gac5y
6734
CHEBI:22154
1,2-dihydroacenaphthylene
C1CC2=CC=CC3=C2C1=CC=C3
Boiling Point 279°C
Color White
Melting Point 95°C
Quantity ZONE
UN Number 3077
Formula Weight 154.21
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.5% (GC)
Chemical Name or Material Acenaphthene Zone Refined (number of passes:30)

EINECSNumber : (4)-0645

RTECSNumber : AB1000000

TSCA : Yes