Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Acenaphthenequinone, 95%

Click to view available options
Quantity:
5g
25g
100g
82-86-0
C12H6O2
182.178
MFCD00003805
AFPRJLBZLPBTPZ-UHFFFAOYSA-N
acenaphthenequinone,acenaphthoquinone,1,2-acenaphthylenedione,acenaphthenedione,1,2-acenaphthenequinone,1,2-acenaphthenedione,acenaphthaquinone,acenaphthylene-1,2-quinone,acenaphthylenequinone,acenaphthodione
6724
CHEBI:15342
acenaphthylene-1,2-dione
C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2
82-86-0
C12H6O2
182.178
MFCD00003805
AFPRJLBZLPBTPZ-UHFFFAOYSA-N
acenaphthenequinone,acenaphthoquinone,1,2-acenaphthylenedione,acenaphthenedione,1,2-acenaphthenequinone,1,2-acenaphthenedione,acenaphthaquinone,acenaphthylene-1,2-quinone,acenaphthylenequinone,acenaphthodione
6724
CHEBI:15342
acenaphthylene-1,2-dione
C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2
Applications
Acenaphthenequinone is used in the preparation of acetylcholinesterase inhibitory agents. It is also used in the preparation of turn-on sensors for cysteine/homocysteine and its application in bioimaging. Further, it is used as an intermediate in manufacturing of dyes and pharmaceuticals. In addition to this, it is used in chemical research as a drug and therapeutic agent.

Solubility
Soluble in chloroform and alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Quantity 100g
Melting Point 253°C to 262°C
Beilstein 879172
Formula Weight 182.18
Percent Purity 95%
Chemical Name or Material Acenaphthenequinone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AB1024500

TSCA : TSCA