Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acenaphthenequinone 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
82-86-0
C12H6O2
182.178
MFCD00003805
AFPRJLBZLPBTPZ-UHFFFAOYSA-N
acenaphthenequinone,acenaphthoquinone,1,2-acenaphthylenedione,acenaphthenedione,1,2-acenaphthenequinone,1,2-acenaphthenedione,acenaphthaquinone,acenaphthylene-1,2-quinone,acenaphthylenequinone,acenaphthodione
6724
CHEBI:15342
acenaphthylene-1,2-dione
C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2
82-86-0
C12H6O2
182.178
MFCD00003805
AFPRJLBZLPBTPZ-UHFFFAOYSA-N
acenaphthenequinone,acenaphthoquinone,1,2-acenaphthylenedione,acenaphthenedione,1,2-acenaphthenequinone,1,2-acenaphthenedione,acenaphthaquinone,acenaphthylene-1,2-quinone,acenaphthylenequinone,acenaphthodione
6724
CHEBI:15342
acenaphthylene-1,2-dione
C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2
Color Undesignated
Melting Point 255°C
Quantity 100g
Formula Weight 182.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Acenaphthenequinone

RTECSNumber : AB1024500

TSCA : Yes