Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acepromazine Maleate, USP, 98-101%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
3598-37-6
C23H26N2O5S
442.53
FQRHOOHLUYHMGG-WLHGVMLRSA-N
(2E)-but-2-enedioic acid; 1-{10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl}ethan-1-one
OC(=O)\C=C\C(O)=O.CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(C)=O
3598-37-6
C23H26N2O5S
442.53
FQRHOOHLUYHMGG-WLHGVMLRSA-N
(2E)-but-2-enedioic acid; 1-{10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl}ethan-1-one
OC(=O)\C=C\C(O)=O.CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(C)=O

A1571, 3598-37-6, C19H22N2OS•C4H4O4

Spectrum™ Acepromazine Maleate, USP is a phenothiazine derivative antipsychotic drug with veterinary sedative and antiemetic properties. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Ignition Residue 0.002
Melting Range 136-139°C
Quantity 5 kg
CAS Max % 1
Percent Purity 98 to 101%