Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ ACES, 0.5M buffer soln., pH 7.5

Click to view available options
Quantity:
100mL
250mL
7365-82-4
C4H10N2O4S
182.194
MFCD00008030
DBXNUXBLKRLWFA-UHFFFAOYSA-N
81832
CHEBI:39062
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
C(CS(=O)(=O)O)NCC(=O)N
7365-82-4
C4H10N2O4S
182.194
MFCD00008030
DBXNUXBLKRLWFA-UHFFFAOYSA-N
81832
CHEBI:39062
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
C(CS(=O)(=O)O)NCC(=O)N
Concentration 0.5M buffer soln.
Physical Form Liquid
pH 7.5
Color Colorless
Quantity 100mL
Chemical Name or Material ACES buffer soln.

TSCA : Yes

Recommended Storage : Keep cold