Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acesulfame Potassium, 99%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
Poly Pail
55589-62-3
C4H4KNO4S
201.24
JLEKLYQXZHJOTQ-UHFFFAOYSA-M
potassium 6-methyl-2,4-dioxo-4H-1,2λ⁶,3-oxathiazin-2-olate
[K+].CC1=CC(=O)N=S([O-])(=O)O1
55589-62-3
C4H4KNO4S
201.24
JLEKLYQXZHJOTQ-UHFFFAOYSA-M
potassium 6-methyl-2,4-dioxo-4H-1,2λ⁶,3-oxathiazin-2-olate
[K+].CC1=CC(=O)N=S([O-])(=O)O1

A2815, 55589-62-3, C4H4KNO4S

Spectrum™ Acesulfame Potassium is a sugar substitute that has no calories and is a white crystalline powder. Thanks to the fact that it is 200 times sweeter than sugar and stable under heat, it is used commercially as a sweetener in baking and products that need to have a long shelf life. It is also used in combination with other sweeteners in carbonated drinks and is typically used as the main sweetening agent in pharmaceutical products.
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 100%
Percent Purity 99%