Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acesulfame Potassium, FCC, 99-101%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
55589-62-3
C4H4KNO4S
201.24
JLEKLYQXZHJOTQ-UHFFFAOYSA-M
potassium 6-methyl-2,4-dioxo-4H-1,2λ⁶,3-oxathiazin-2-olate
[K+].CC1=CC(=O)N=S([O-])(=O)O1
55589-62-3
C4H4KNO4S
201.24
JLEKLYQXZHJOTQ-UHFFFAOYSA-M
potassium 6-methyl-2,4-dioxo-4H-1,2λ⁶,3-oxathiazin-2-olate
[K+].CC1=CC(=O)N=S([O-])(=O)O1

A3996, 55589-62-3, C4H4KNO4S

Spectrum™ Acesulfame Potassium, FCC is also known as acesulfame K or Ace K, and is a calorie-free sugar substitute. This artificial sweetener is 200 times sweeter than sucrose (common sugar), as sweet as aspartame, about two-thirds as sweet as saccharin, and one-third as sweet as sucralose. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.01
Quantity 250 g
CAS Max % 1
Percent Purity 99 to 101%