Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals

Catalog Number AAL1226518
Click to view available options
Quantity:
50 g
58052-80-5
C6H18N3
132.23
MFCD00149559
MZSSRMMSFLVKPK-UHFFFAOYSA-Q
2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt
2723814
2,4,6-trimethyl-1,3,5-triazinane;trihydrate
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1
58052-80-5
C6H18N3
132.23
MFCD00149559
MZSSRMMSFLVKPK-UHFFFAOYSA-Q
2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt
2723814
2,4,6-trimethyl-1,3,5-triazinane;trihydrate
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1

 

Acetaldehyde ammonia trimer is used as scavengers for sulfhydryl compounds in natural gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde ammonia trimer is used as scavengers for sulfhydryl compounds in natural gas.

Solubility
Soluble in water.

Notes
Desiccate at 4°C. Store away from air, moisture and oxidizing agents
TRUSTED_SUSTAINABILITY
Melting Point 93°C to 96°C
Density 0.852
Boiling Point 110°C (decomposition)
Quantity 50 g
UN Number UN1841
Beilstein 3910586
Sensitivity Hygroscopic
Merck Index 14,40
Solubility Information Soluble in water.
Formula Weight 183.25
Percent Purity 98%
Chemical Name or Material Acetaldehyde ammonia trimer
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ACETALDEHYDE AMMONIA
EINECSNumber 200-868-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit