Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetaldehyde diethyl acetal, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.
Solubility
Soluble in water (46 g/L at 20°C).
Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with oxidizing agents.
Acetaldehyde diethyl acetal is used as a flavoring agent to provide fruit, nut, rum, and whiskey flavors.
Solubility
Soluble in water (46 g/L at 20°C).
Notes
Keep container tightly sealed. Store in refrigerated condition. Incompatible with oxidizing agents.
Specifications
Melting Point | -100°C |
Density | 0.831 |
Boiling Point | 101°C to 102°C |
Flash Point | −21°C (−6°F) |
Odor | Pleasant |
Refractive Index | 1.381 |
Quantity | 100 g |
UN Number | UN1088 |
Beilstein | 1098310 |
Merck Index | 14,38 |
Show More |
Safety and Handling
Hazard Category | H225-H315-H319 |
Hazard Statement | GHS H Statement H225-H315-H319 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETAL |
EINECSNumber | 203-310-6 |
RTECSNumber | AB2800000 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction