Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde ethyl propargyl acetal, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
18669-04-0
C7H12O2
128.17
MFCD00066344
QKBBTQJLUGADEG-UHFFFAOYSA-N
3-1-ethoxyethoxy prop-1-yne,acetaldehyde ethyl propargyl acetal,1-propyne, 3-1-ethoxyethoxy,1-ethoxy-1-prop-2-yn-1-yloxy ethane,3-1-ethoxyethoxy propyne,1-ethoxyethoxy-2-propyne,1-1-ethoxyethoxy-2propyne,1-1-ethoxyethoxy-2-propyne,5-methyl-4,6-dioxa-1-octyne,1-propyne,3-1-ethoxyethoxy
86774
3-(1-ethoxyethoxy)prop-1-yne
CCOC(C)OCC#C
18669-04-0
C7H12O2
128.17
MFCD00066344
QKBBTQJLUGADEG-UHFFFAOYSA-N
3-1-ethoxyethoxy prop-1-yne,acetaldehyde ethyl propargyl acetal,1-propyne, 3-1-ethoxyethoxy,1-ethoxy-1-prop-2-yn-1-yloxy ethane,3-1-ethoxyethoxy propyne,1-ethoxyethoxy-2-propyne,1-1-ethoxyethoxy-2propyne,1-1-ethoxyethoxy-2-propyne,5-methyl-4,6-dioxa-1-octyne,1-propyne,3-1-ethoxyethoxy
86774
3-(1-ethoxyethoxy)prop-1-yne
CCOC(C)OCC#C
Refractive Index 1.4150 to 1.417
Density 0.8930g/mL
Boiling Point 141°C to 142°C
Specific Gravity 0.893
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 128.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Acetaldehyde ethyl propargyl acetal
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 242-490-