Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde, Reagent, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
75-07-0
C2H4O
44.05
IKHGUXGNUITLKF-UHFFFAOYSA-N
acetaldehyde
CC=O
75-07-0
C2H4O
44.05
IKHGUXGNUITLKF-UHFFFAOYSA-N
acetaldehyde
CC=O

A2147, 75-07-0, C2H4O

Spectrum™ Acetaldehyde, Reagent, ACS is commercially used as a precursor to acetic acid. In the laboratory it is a reagent used for the detection of primary and secondary amines. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Packaging Amber Glass Bottle
Residue after Evaporation 5e-005
Quantity 1 L
CAS Max % 1
UN Number UN1089
Percent Purity 99.5%