Acetamide (Crystalline/Certified), Fisher Chemical

Packaging Glass Bottle
Quantity 250g

 View more versions of this product

Catalog No. A4-250

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 60-35-5
    Physical Form Solid
    Molecular Formula C2H5NO
    Linear Formula CH3CONH2
    Formula Weight 59.07
    MDL Number MFCD00008023
    Synonym Acetic Acid Amide
    Chemical Name or Material Acetamide
    Grade Certified Grade

    Specifications

    Formula Weight 59.07
    Boiling Point 221°C
    Melting Point 79°C
    Quantity 250g
    Physical Form Solid
    Color White
    Density 1.16g/mL
    Identification Pass Test
    Heavy Metals (as Pb) 0.001% max.
    Acetate 1% max.
    Ignition Residue 0.010% max.
    Molecular Formula C2H5NO
    Linear Formula CH3CONH2
    MDL Number MFCD00008023
    CAS 60-35-5
    ChemAlert Storage Symbol Gray
    Packaging Glass Bottle
    Synonym Acetic Acid Amide
    Sulfate 0.002% max.
    Chloride 0.002% max.
    Chemical Name or Material Acetamide
    Grade Certified Grade
     

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit